VWR® Tubing, PE
Fournisseur: VWR International
Certificats
A propos de cet article
Polyethylene tubing is engineered to provide over three times the impact and twice the tensile strength of traditional low-density polyethylene.
- Clear polyethylene construction
- Heat sealable
- Superior tensile strength to traditional LDPE
Clear polyethylene tubing is heat sealable. Wound on 7.6 cm (3") core. Core plug has a 2.5 cm (1") diameter hole. Manufactured with 100% renewable energy.
Customers Who Bought This Also Bought