VWR® Reclosable Parts Bags
Fournisseur: VWR International
CE Compliant
Certificats
A propos de cet article
These bags have been chosen for their ability to rival any reclosable bag at economical prices.
- Full gauge
- Exceptional clarity and quality
- Hanging hole
- Prepared writing area
These reclosable bags are full gauge and offer exceptional clarity and quality. They feature a large white writing area that accepts either marker or pen and a single hang hole above the zipper closure for a secure hang. These tough bags employ a reinforced side-welded zipper to lock in single smooth motion. Made of 100% virgin Low Density Polyethylene (LDPE) resin.
Certifications: Meet FDA/USDA specifications.
Customers Who Bought This Also Bought