VWR® Poly Bags, Gussetted
Fournisseur: VWR International
CE Compliant
Certificats
A propos de cet article
Each of VWR® imported gusseted poly bags are made of 100% virgin Low Density Polyethylene (LDPE) resin.
- Tubular construction and durable bottom seal
- Outstanding vapor and moisture barrel
- Twice the tensile strength of traditional LDPE bags
- Made with 100% renewable energy
Side gussets expand easily to fit over bulky or irregular size objects. Features superior clarity for showing the quality of the customer's products. 100 packed. All listed gauges are nominal. Can be heat sealed, tied, stapled or taped shut.
Certifications: Meet FDA/USDA specifications.
Customers Who Bought This Also Bought