Barnstead Air Filter and Water Seal for Distilled Water Storage Tanks, Thermo Scientific
# de catalogue 26142-000
Fournisseur: Thermo Fisher Scientific
Spécifications
- Description:Air Filter and Water Seal
- Cat. no.:26142-000
- Supplier No.:H3111
Spécifications
A propos de cet article
Water seal for 1 to 10 gallon per hour input.
- Protects stored distilled water against airborne impurities
- Prevents organic vapors, alkali, and acid gases from entering the tank
Effectively filters out all dust and particles from air with 100% efficiency, even with particles as small as 0.2 µm. Under normal conditions, one cartridge lasts 60 days, or until 1000 gallons of distilled water have been drawn from the tank. A simple screw cap makes changing of filters quick and easy.
Ordering information: Supplied with seal and fittings for metal tank, including water seal chamber, fittings for connection to tank, sealing chamber with drain cock, and roll of special tape for sealing tank cover.
Customers Who Bought This Also Bought