Barnstead™ Single Filter Holder, Economy, Thermo Scientific
# de catalogue 26303-020
Fournisseur: Thermo Fisher Scientific
Spécifications
- Description:Economy Filter Holder
- Max. inlet pressure:100 psi
- Cat. no.:26303-020
- Supplier No.:15840
Spécifications
A propos de cet article
Single filter holder accepts Thermo Scientific double open-end prefilters or final filters.
- Reinforced PP construction
- Features push-button air bleed and Buna-N, O-rings
- Inlet/Outlet connections are 3/4" NPT
Customers Who Bought This Also Bought