Barnstead Bantam™ Deionizer Holders, Thermo Scientific
Supplier: Thermo Fisher Scientific
Certificates
About this item
These holders connects directly to raw water line and delivers demineralized water in continuous flow at atmospheric pressure.
- Simple thumbwheel arrangement permits cartridge change without tools
- Direct reading meter indicates purity of effluent at all times
- Pedestal is removable if wall-mounting is preferred
- Shows when cartridge needs replacing
Meter calibrated at 0-19 ppm (as NaCl) and in ohms resistance from 25000 to 18000000 ohms-cm.
Delivery: Supplied with 0.9 m (3') plastic tube to connect to water line, inlet flow adjustment valve, drain valve, on-off switch, cord and plug. Cartidges not included.
Customers Who Bought This Also Bought