Specifications
- Capacity:237 ml (8 oz.)
- Description:Sample Container with Pour Spout
- Cat. no.:15704-050
Specifications
About this item
Clear, graduated polystyrene container features a pour spout and a paper lid.
- Pour spout
- Clear siding
- Graduated markings
- Polystyrene construction
Graduations are marked from 10 to 200cc in 10cc intervals and from 1/2 to 61/2oz. in 1/2 oz. intervals. Height: 9.1 cm (39/16"). Top dia.: 7.1 cm (213/16"). Top dia. with spout: 7.5 cm (215/16"). Bottom dia.: 5.1 cm (2").