VWR® Talon® Hosecock Extension Clamp
Catalog # 21716-110
Supplier: VWR International
Product available on GSA Advantage®
Specifications
- Cat. no.:21716-110
- Maximum Grip Size:17 mm (²¹⁄₃₂")
Specifications
About this item
Similar to regular hosecock but with a 145 mm (523/32") bottom-mounted extension rod for attachment to VWR® Talon® Lab Frame rods.
- Delivers precise flow control
- Blind mounting hole eliminates potential damage to tubing
- Easy adjustment and location of tubing with automatic offset
- Secure retention of tubing
Blind mounting hole of extension rod to hosecock eliminates potential damage to tubing. Automatic offset to allow easy adjustment and location of tubing. Clamp features a fine machined adjustment screw with oversized head for accurate regulation of fluid flow. Tubing retainer screw on side of clamp can be raised to allow easy positioning of tubing. When screwed back in place, retainer screw keeps tubing from disengaging under varying fluid pressures. 90° holders sold separately (see 21572-501 and 21572-556).
Customers Who Bought This Also Bought