SP Bel-Art Spring Style Tubing Clamps, Bel-Art Products, a part of SP
Fournisseur: Bel-Art Products, a Part of SP
Certificats
A propos de cet article
Perfect for all flexible tubing, this positive-action, self-closing clamp opens by squeezing the handles together.
- Plastic and stainless steel construction
- Self-closing clamp design
- Easy to use
- Suitable for use with all flexible tubing
Made of chemical and stain resistant Acetal plastic and stainless steel.
Customers Who Bought This Also Bought