Space-Saver Flammable Liquids Storage Cabinets, Horizontal, Eagle Manufacturing
Fournisseur: Eagle Manufacturing
Certificats
A propos de cet article
Keep flammable liquids near work areas in these small storage cabinets, or stack them on top of 113.6 L (30 gal.) or 170.3 L (45 gal.) storage cabinets to add space where needed.
- Three-point key lock on doors
- 18-gauge steel with double-wall design
Like their full-size counterparts, these units are constructed of tough, has 3.8 cm (1¹/₂") air space between walls. Finished in a caution yellow powder-coat paint with 'FLAMMABLE–KEEP FIRE AWAY' in red lettering on the front. Label is in three languages (English, Spanish, French). Additional features include grounding attachments, and 5.1 cm (2") raised leakproof door sills.
Certifications: Cabinets meet OSHA and NFPA Code 30 specifications. Self-closing models meet UFC 79.
Customers Who Bought This Also Bought