Flammable Liquids Safety Storage Cabinets, Eagle Manufacturing
Fournisseur: Eagle Manufacturing
Certificats
A propos de cet article
Storage cabinets constructed of 18-gauge steel with a caution yellow powder-coat finish with “FLAMMABLE–KEEP FIRE AWAY” in red lettering on the front.
- Label is in three languages (English, Spanish, French)
- The sides, top, bottom and doors are double-walled
- 3.8 cm (1¹/₂") air space between walls
Cabinets with self-closing doors are equipped with a fusible link that automatically closes the door if the link melts due to fire.
Vents have 5 cm (2") pipe-thread fittings and are equipped with a fire baffle and cap. Cabinets are equipped with grounding attachments, three-point key lock on doors, 5 cm (2") raised, leakproof door sills, and four leveling feet. Each adjustable shelf is supported by four brackets.
Certifications: Conform to OSHA flammable liquid storage requirements and NFPA Code 30. Models with a capacity of 113.6 L (30 gal.) to 227.1 L (60 gal.) are FM approved.
Customers Who Bought This Also Bought