Corning® Aseptic Transfer Caps, Corning
Fournisseur: Corning
Certificats
A propos de cet article
Disposable transfer caps are for use with plastic Erlenmeyer flasks (see 29152-146 series)
Internal end of liquid transfer port rigid polypropylene dip tube angled to reach bottom of flask; external side has OD C-Flex tubing closed with female luer sealing cap.
Vent filter is 50 mm (2") in diameter, with a 0.2 µm pore PVDF membrane and barbed fitting. Internal end of liquid transfer port rigid polypropylene dip tube angled to reach bottom of flask; external side has OD C-Flex tubing closed with female luer sealing cap.
Delivery: Transfer caps are supplied with a Sterility Assurance Level (SAL) of 10-6, and are double bagged.
Customers Who Bought This Also Bought