KONTES® West Condensers, Medium Length, Kimble Chase
Fournisseur: DWK Life Sciences (KIMBLE)
Certificats
A propos de cet article
For refluxing and distillation assemblies.
Condensate is not held up with this design. #1 hose connections.
Customers Who Bought This Also Bought