Squeeze-O-Matic Burets, AMK Glass
Fournisseur: AMK GLASS INC
Certificats
A propos de cet article
Automatic, self-zeroing burets are ideal for first time users, or as a student learning tool
Glass stopcocks are the best selection for use with acid solutions, where stopcock grease can be applied. When properly greased, glass stopcocks provide excellent leak-free service. The PTFE stopcocks are for use with alkaline solutions to avoid "freezing."
Burets feature a durable white ceramic enamel graduation scale. Squeezable bottles are made of low-density polyethylene (LDPE). Bottles include a detachable base stand for stability. Squeezing the bottle forces the titrant into the buret. The overfill returns to the bottle when the pressure is released.
Customers Who Bought This Also Bought