Specifications
- No. of wells:96
- Skirt:Non-skirted
- Well Color:Clear
- Well Volume:0,30 ml
- Environmentally Preferable:Low Manufacturing Impact
- Environmental certification:I-RECs, ISO 14001 - Reynosa
- Cat. no.:47734-766
- Supplier No.:PCR-96-MB-C
Specifications
About this item
These plates are made of 99.9% pure virgin polypropylene and specifically designed for PCR applications
- Max. RCF 4000 g
Optional AxyMat™ silicone sealing mat (10011-130) is autoclavable and freezable. Mats are pierceable and self-sealing to prevent escape of evaporated aerosols and pathogens during normal use or storage. Alphanumeric grid system corresponds to 96-well plates.
They feature elevated chimney wells that are slightly taller and narrower than typical chimney wells, making these plates ideal for use with Amersham Biosciences MegaBACE™ DNA analysis systems. Plates are ideal for thermal transfer applications because their ultra-thin walls accelerate heat transfer to provide shorter cycles. Certified by lot number to be RNase-, DNase-, and pyrogen-free.