Axygen® 96-Well PCR Plates Scientific
Fournisseur: Corning
Certificats
A propos de cet article
PP plates, silicone mats. Designed to fit 0.2 ml thermal cycler blocks.
- Ultra-thin walls of uniform thickness accelerate heat transfer, leading to shorter cycle times and more consistent results
- Certified free from DNase, RNase and endotoxin for highest sample integrity
- AxyMat™ sealing mats autoclavable and reusable
Volume with AxyMat™: 240 μl
Compatible with (in addition to standard thermal cycling): ABI Fast PCR 5700, 7000, 7700, BioRad iCycler, MyiQ, iQ5, ABI sequencers.
Spécifications
- Catalog No:
- 47749-876
- 10011-220
- 10011-222
- 10011-248
- 10011-228
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- I-RECs, ISO 14001 - Reynosa
- I-RECs, ISO 14001 - Reynosa
- I-RECs, ISO 14001 - Reynosa