A propos de cet article
These plates are made of 99.9% pure virgin polypropylene and are specifically designed for PCR and thermal transfer applications
Optional AxyMat™ silicone sealing mats are autoclavable and freezable. The standard AxyMat™ is pierceable and self-sealing to prevent escape of evaporated aerosols and pathogens during normal use or storage. It also features an alphanumeric grid system that corresponds to 96-well plates. The AxyMat™ septa mat features individual septa to allow easy penetration and to prevent sample contamination and evaporation. It is designed specifically for use with the ABI PRISM® 3100 system.
- Max. RCF 4000 g
These plates are made of 99.9% pure virgin polypropylene and are specifically designed for PCR and thermal transfer applications
Optional AxyMat™ silicone sealing mats are autoclavable and freezable. The standard AxyMat™ is pierceable and self-sealing to prevent escape of evaporated aerosols and pathogens during normal use or storage. It also features an alphanumeric grid system that corresponds to 96-well plates. The AxyMat™ septa mat features individual septa to allow easy penetration and to prevent sample contamination and evaporation. It is designed specifically for use with the ABI PRISM® 3100 system.
Half-skirt design with A12 corner notch fits the Applied Biosystems ABI PRISM® 3100 Genetic Analyzer. Ultra-thin walls accelerate heat transfer to provide shorter cycles. All plates are certified by lot number to be RNase-, DNase-, and pyrogen-free.
Spécifications
- Catalog No:
- 47745-424
- 47743-953
- 47745-430
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Working volume:
- 300 µL
- 300 µL
- Environmental certification:
- I-RECs, ISO 14001 - Reynosa
- I-RECs, ISO 14001 - Reynosa
- I-RECs, ISO 14001 - Reynosa