ContactUsLinkComponent
CLP–Semi-Volatiles DFTPP Tuning Compound, Restek
CLP–Semi-Volatiles DFTPP Tuning Compound, Restek
# de catalogue RK31001
Fournisseur:  Restek
Numéro CAS:  
CLP-Semi-Volatiles DFTPP Tuning Compound
CLP–Semi-Volatiles DFTPP Tuning Compound, Restek
# de catalogue RK31001
Fournisseur:  Restek
Numéro CAS:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Description:
    Tuning Compound
  • Cat. no.:
    RK31001
  • Supplier No.:
    31001
Spécifications
A propos de cet article
Standard is packaged in methylene chloride at a concentration of 2500 µg/mL in a 1 mL ampoule.
  • Mixture contains Decafluorotriphenylphosphine (DFTPP)
Each standard includes an ampoule breaker, deactivated amber screw-top vial, extra product label, Certificate of Analysis, and Material Safety Data Sheet (MSDS).
Customers Who Bought This Also Bought