VWR® Graduated Urine Centrifuge Tubes, Polystyrene
Supplier: VWR International
Certificates
About this item
These leakproof disposable tubes are made of USP VI PS resin and feature molded graduations.
- Molded graduations stand out distinctly on these optically clear polystyrene tubes
- RNase-, DNase-, and Endotoxin-Free
- Not autoclavable
They are colorless and transparent which is perfect for biological reaction experiments.
Certifications: Manufactured in a class 100 000 room environment. Manufactured under ISO13485:2016 and ISO9001:2015 quality management. Meet the requirements of RoHS.
Specifications
- Catalog No:
- 21008-000
- 21008-022
- 21008-035
- Environmentally Preferable:
- Sustainable Packaging, Low Manufacturing Impact
- Sustainable Packaging, Low Manufacturing Impact
- Sustainable Packaging, Low Manufacturing Impact