A propos de cet article
Abbe refractometers are available in a variety of models for both high and low refractive index measurement, as well as high temperature and precise measurement needs
The NAR-3T has been developed to give improved measurement accuracy and ease of use through an advanced optical system, larger scale, high-intensity lamp, and double control knob. And for the measurement of materials with a high refractive index, model NAR-4T is available.
NAR-1T LIQUID and NAR-1T SOLID designs has been developed for the measurement of liquid and solid samples respectively. And NAR-2T designed for use with compounds which require measurement at high temperatures.
They are designed to give refractive indices of the D line of solids and liquids without using a sodium lamp.
Spécifications
- Catalog No:
- 47752-794
- 97015-542
- 47752-796
- 47752-782
- 97015-880
- 47752-780
- 47752-798
- 97015-544
- Power consumption:
- 5VA
- 5VA
- 5VA
- 5VA
- 5VA
- 5VA
- 5VA
- 5VA
- W×D×H:
- 8W×10D×11H cm (33/16×315/16×45/16")
- 8W×10D×11H cm (33/16×315/16×45/16")
- 8W×10D×11H cm (33/16×315/16×45/16")
- 8W×10D×11H cm (33/16×315/16×45/16")
- 8W×10D×11H cm (33/16×315/16×45/16")
- 8W×10D×11H cm (33/16×315/16×45/16")
- 8W×10D×11H cm (33/16×315/16×45/16")
- 8W×10D×11H cm (33/16×315/16×45/16")