SP Bel-Art ProCulture Stak-a-Tray System, Bel-Art Products, a part of SP
Fournisseur: Bel-Art Products, a Part of SP
Certificats
A propos de cet article
Modular rack and tray system maximizes storage for Petri dishes and cell culture flasks in incubators, cold rooms or on the bench.
- Stainless steel construction
- Modular design allows customization and maximized storage
- Autoclavable
Each rack has 4 tiers
Each tier accommodates one large or two small trays. All parts are stainless steel and autoclavable at 121ºC (250ºF).
Stack up to five racks for 20 levels of storage. Perforated trays allow air flow and slide easily in the rack.
Ordering information: Racks and trays are sold separately.
Customers Who Bought This Also Bought