Single Drum Spill Containment Station, Eagle Manufacturing
Fournisseur: Eagle Manufacturing
Certificats
A propos de cet article
For secondary spill containment of hazardous materials.
Blow-molded high-density polyethylene unit with 321.8L (85gal.) spill capacity is suitable for 208.2L (55gal.) drums or smaller. Use as a collection or dispensing center. Chemically-resistant to most materials. Forkliftable. Nestable for easy storage.
Customers Who Bought This Also Bought