Double Drum Secondary Containment Workstation, Eagle Manufacturing
Fournisseur: Eagle Manufacturing
Certificats
A propos de cet article
For secure indoor or outdoor drum storage.
Heavy-duty polyethylene unit in safety yellow holds two 208.2L (55gal.) drums. Use as a collection station or dispensing center for most hazardous chemicals. Secondary spill capacity of 454L (120gal.). Forkliftable. Comes complete with locks.
Meets EPA requirement 40 CFR 264.175.
Customers Who Bought This Also Bought