Stackable and Wall-Mounted Flammables Safety Cabinets, SECURALL®
Fournisseur: Securall Cabinets
Certificats
A propos de cet article
Mount these cabinets to the wall or stack them for convenient flammable liquid storage without taking valuable workspace.
- Each cabinet is supplied with brackets for easy mounting
- Self-close cabinets feature a fusible link that melts at 71 °C (165 °F), allowing the doors to automatically close and latch
- All cabinets have a large 'FLAMMABLE – KEEP FIRE AWAY' label
Wall-mounted cabinets are made of double-wall 18-gauge galvanized steel and include five adjustable 18-gauge galvanized steel shelves, full-length heavy-gauge piano hinges, flush-mounted locking handle, and a three-point self-latching closure.
Certifications: Cabinets meet NFPA Code 30 specifications. FM approved.
Spécifications
- Catalog No:
- 15001-712
- 14227-658
- 15001-710
- 15001-708
- 14227-660
- 15001-714
- Environmentally Preferable:
- Sustainable Materials
- Sustainable Materials
- Sustainable Materials
- Sustainable Materials
- Sustainable Materials
- Sustainable Materials
- Environmental certification:
- SMaRT Certified
- SMaRT Certified
- SMaRT Certified
- SMaRT Certified
- SMaRT Certified
- SMaRT Certified