VWR® Spinring® Magnetic Stir Bars
Fournisseur: VWR International
Certificats
A propos de cet article
Octagonal stirring bar is contained within a removable stabilizing ring that centers it in a container and creates a deep vortex without splashing, chattering, or spinout.
- Removable stabilizing ring keeps bar centered
- Virtually eliminates spinout
- No chattering noise
- Autoclavable
Alnico magnet encapsulated in FDA grade Teflon® PTFE is durable and inert.
Customers Who Bought This Also Bought