Glove Box Gloves, Labconco
Fournisseur: Labconco
Certificats
A propos de cet article
Neoprene straight and accordion sleeves can be used with hand-specific neoprene or latex hands.
- Hypalon® gloves are resistant to abrasion, ozone, and oxidizing chemicals
Neoprene, butyl, and Hypalon® gloves are one-piece, seamless, and hand-specific. Butyl gloves provide high impermeability to water vapor, gases, and toxic chemicals, and are resistant to oxygenated solvents and most oxidizing chemicals.
Ordering information: Sleeves come complete with O-rings and spring. Neoprene, butyl, and Hypalon® gloves come with O-rings.
Spécifications
- Catalog No:
- 32948-205
- 15202-519
- 32948-203
- 32948-154
- 32948-019
- 32948-096
- Ambidextrous/Hand-specific:
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Latex/Latex-free:
Spécifications
Customers Who Bought This Also Bought