VWR® Spinbar® Octagon Stir Bars, Color-Coded and Assorted
# de catalogue 58948-074
Fournisseur: VWR International
Spécifications
- Coating:PTFE
- Description:Stir Bars
- Cat. no.:58948-074
- Shape:Octagon
Spécifications
A propos de cet article
An assortment of 24 PTFE resin-coated, magnetic stir bars color-coded in red, yellow, and blue.
- Color-coded
- PTFE resin coating
- Interrupted profile offers added turbulence
- Most common stir bar shape
Available in red, yellow, or blue. Packaged in a plastic, compartmentalized Spinbox™.
Ordering information: Includes one of each color in each size: 12.7x3.2mm (1/2x1/8"), 12.7x7.9mm (1/2x5/16"), 15.9x7.9mm (5/8x5/16"), 22.2x7.9mm (7/8x5/16"), 25.4x7.9mm (1x5/16"), 38.1x7.9mm (11/2x5/16"), 50.8x7.9mm (2x5/16"), and 76.2x12.7mm (3x1/2").
Customers Who Bought This Also Bought