About this item
Powerful magnetic stirrers with precise speed control offer repeatable performance.
- Stir up to 9 vessels at once
- Microprocessor speed control
- Synchronized, uniform mixing
- Low-profile design
- Now with Best-in-Class, Exclusive 5-Year Warranty
Synchronized operation ensures uniform mixing at each position
Model 650 standard series offers an analog speed control knob with dial markings. Model 750 advanced series offers an LED display for digital speed control, adjustable in 10rpm increments.
Ramping feature slowly brings sample to set speed to avoid splashing, improve magnetic coupling, and provide excellent low-speed stirring. Large top plate with position indicators is easy to clean and provides a white reflective surface. Low-profile design takes up less space and easily fits into fume hoods. Spill-resistant housing channels fluids away from internal components. Cast metal base offers durability and added stability.
Specifications
- Catalog No:
- 10027-068
- 10027-070
- 12621-026
- 12621-018
- 12621-042
- 12621-022
- 12621-046
- 12621-034
- 10027-076
- 10027-078
- 12621-038
- Speed range:
- 60 - 1400 rpm
- 60 - 1400 rpm
- 60 - 1400 rpm
- 60 - 1400 rpm
- 60 - 1400 rpm
- 60 - 1400 rpm
- 60 - 1400 rpm
- 60 - 1400 rpm
- 60 - 1400 rpm
- 60 - 1400 rpm
- 60 - 1400 rpm
- Plate dimensions:
- 27.9W×31.8D cm (11×12½")
- 27.9W×31.8D cm (11×12½")
- 27.9W×31.8D cm (11×12½")
- 27.9W×31.8D cm (11×12½")
- 27.9W×31.8D cm (11×12½")
- 27.9W×31.8D cm (11×12½")
- 27.9W×31.8D cm (11×12½")
- 27.9W×31.8D cm (11×12½")
- 27.9W×31.8D cm (11×12½")
- 27.9W×31.8D cm (11×12½")
- 27.9W×31.8D cm (11×12½")
- W×D×H:
- 27.9×43.2×10.2 cm (11×17×4")
- 27.9×43.2×10.2 cm (11×17×4")
- 27.9×43.2×10.2 cm (11×17×4")
- 27.9×43.2×10.2 cm (11×17×4")
- 27.9×43.2×10.2 cm (11×17×4")
- 27.9×43.2×10.2 cm (11×17×4")
- 27.9×43.2×10.2 cm (11×17×4")
- 27.9×43.2×10.2 cm (11×17×4")
- 27.9×43.2×10.2 cm (11×17×4")
- 27.9×43.2×10.2 cm (11×17×4")
- 27.9×43.2×10.2 cm (11×17×4")
- Electrical:
- 120 V, 50/60 Hz, 1.3 A, 85 W
- 120 V, 50/60 Hz, 1.3 A, 85 W
- 120 V, 50/60 Hz, 1.3 A, 85 W
- 120 V, 50/60 Hz, 1.3 A, 85 W
- 120 V, 50/60 Hz, 1.3 A, 85 W
- 120 V, 50/60 Hz, 1.3 A, 85 W
- 120 V, 50/60 Hz, 1.3 A, 85 W
- 120 V, 50/60 Hz, 1.3 A, 85 W
- 120 V, 50/60 Hz, 1.3 A, 85 W
- 120 V, 50/60 Hz, 1.3 A, 85 W
- 120 V, 50/60 Hz, 1.3 A, 85 W