KIMAX® Glass Stirring Rods, Kimble Chase
Fournisseur: DWK Life Sciences (KIMBLE)
Certificats
Certificats
A propos de cet article
With rounded annealed ends
Designed from Federal Specification NNN-R-560, Type I requirements.
Customers Who Bought This Also Bought
Nalgene® VERSI-DRY® Lab Table Soakers, Thermo Scientific
Fournisseur: Thermo Fisher Scientific
1 098,06 $
VACUETTE® Sodium Citrate Coagulation Tubes, Greiner Bio-One
Fournisseur: Greiner Bio-One
18,45 $ to 442,53 $
Ethanol denatured ACS (denatured with 5% isopropanol and 5% methanol), VWR Chemicals BDH®
Fournisseur: VWR International
411,94 $ to 1 303,38 $
MiniCollect* Tube K2E K2Edta Non sterile Lavender Cap 0.25/0.5 Ml PK500
Fournisseur: Greiner Bio-One
656,98 $ to 2 627,82 $
VWR® Absorbent Underpads with Waterproof Moisture Barrier
Fournisseur: VWR International
66,85 $ to 318,03 $
VWR® Laboratory Bench and Table Protectors with Leakproof Moisture Barrier
Fournisseur: VWR International
359,30 $
Diversey™ Oxivir® Tb Disinfectant Solution, Antylia Scientific
Fournisseur: Argos Technologies
212,65 $
Osmium tetraoxide ≥99.95%, crystalline, highest purity
Fournisseur: Electron Microscopy Sciences
91,43 $