Multi-Wavelength Abbe Refractometers, ATAGO®
Fournisseur: Atago
Certificats
A propos de cet article
These refractometers measure refractive index or Abbe number ([nu]d or [nu]e) at different wavelengths ranging from 450–1100nm for model DR-M2 and DR-M4 and from 450–1550nm for model DR-M2/1550 and DR-M4/1550
For measurement at wavelengths ranging from 681–1100nm using models DR-M2 and DR-M4, the optional near infrared ray viewer is required. Contact your VWR representative for information.
The units display measurement results on the display.
Ordering information: All units are supplied with a one-year manufacturer’s warranty. Refractometers DR-M2 and DR-M4 are supplied with a test piece (refractive index point), 4mL of monobromonaphthalene, an Allen wrench, a lighting glass, one 589nm interference filter, one 546nm interference filter, 10 tube bands, a spare bulb, and an instruction manual. The DR-M2/1550 and DR-M4/1550 units come with a near infrared ray viewer, mounting adapter and set screw, monochromatic light source device, a test piece (refractive index point), 4mL of monobromonaphthalene, an Allen wrench, a 589nm interference filter, 10 tube bands, and an instruction manual. DR-M4 and DR-M4/1550 also include 4mL of methylene iodide-containing sulfur solution. A calibration certificate and compatible digital printer are available for each unit. Contact your VWR representative for information.
Spécifications
- Catalog No:
- 47752-790
- 47752-792
- 47752-786
- 47752-788
- Model:
- DR-M4
- DR-M4/1550
- DR-M2
- DR-M2/1550
- Measurement range:
- 1.5219–1.9220 nD for 450 nm; 1.4700–1.8700 nD for 589 nm; 1.4545–1.8544 nD for 680 nm; 1.4260–1.8259 nD for 1100 nm
- 1.5219–1.9220 nD for 450 nm; 1.4700–1.8700 nD for 589 nm; 1.4545–1.8544 nD for 680 nm; 1.4260–1.8259 nD for 1100 nm; 1.4138–1.8136 nD for 1550 nm
- 1.3278–1.7379 nD for 450 nm; 1.3000–1.7100 nD for 589 nm; 1.2912–1.7011 nD for 680 nm; 1.2746–1.6843 nD for 1100 nm
- 1.3278–1.7379 nD for 450 nm; 1.3000–1.7100 nD for 589 nm; 1.2912–1.7011 nD for 680 nm; 1.2743–1.6840 nD for 1100 nm; 1.2662–1.6759 nD for 1550 nm
- Measurement range (°Brix):
- 1.5219–1.9220 nD for 450 nm; 1.4700–1.8700 nD for 589 nm; 1.4545–1.8544 nD for 680 nm; 1.4260–1.8259 nD for 1100 nm
- 1.5219–1.9220 nD for 450 nm; 1.4700–1.8700 nD for 589 nm; 1.4545–1.8544 nD for 680 nm; 1.4260–1.8259 nD for 1100 nm; 1.4138–1.8136 nD for 1550 nm
- 1.3278–1.7379 nD for 450 nm; 1.3000–1.7100 nD for 589 nm; 1.2912–1.7011 nD for 680 nm; 1.2746–1.6843 nD for 1100 nm
- 1.3278–1.7379 nD for 450 nm; 1.3000–1.7100 nD for 589 nm; 1.2912–1.7011 nD for 680 nm; 1.2743–1.6840 nD for 1100 nm; 1.2662–1.6759 nD for 1550 nm
- Measurement temperature range:
- 5 to 50°C (41 to 122°F)
- 5 to 50°C (41 to 122°F)
- 5 to 50°C (41 to 122°F)
- 5 to 50°C (41 to 122°F)
- Min. indication:
- 0.0001 in refractive index, 0.1 in Abbe number
- 0.0001 in refractive index, 0.1 in Abbe number
- 0.0001 in refractive index, 0.1 in Abbe number
- 0.0001 in refractive index, 0.1 in Abbe number
- Electrical:
- 100–240V, 50/60Hz
- 100–240V, 50/60Hz
- 100–240V, 50/60Hz
- 100–240V, 50/60Hz
- Dimensions:
- 13W x 29D x 31H cm (51/8 x 117/16 x 121/4") for Refractometer; 15W x 33D x 11H cm (515/16 x 13 x 45/16") for Light Source Unit
- 13W x 29D x 31H cm (51/8 x 117/16 x 121/4") for Refractometer; 15W x 33D x 11H cm (515/16 x 13 x 45/16") for Light Source Unit
- 13W x 29D x 31H cm (51/8 x 117/16 x 121/4") for Refractometer; 15W x 33D x 11H cm (515/16 x 13 x 45/16") for Light Source Unit
- 13W x 29D x 31H cm (51/8 x 117/16 x 121/4") for Refractometer; 15W x 33D x 11H cm (515/16 x 13 x 45/16") for Light Source Unit