About this item
Absolute high-precision thermometers provide the exact readings necessary for critical requirements, quality control checks, and routine measurements.
- Traceable
- Temperature in °C and °F
- Stainless steel probe
- Continuous visual and audio alerts of temperature changes
- One-button self-calibration
These thermometers are ideal for monitoring temperatures in water baths, cuvettes, gas systems, reagents, standards solutions, ovens, incubators, pharmaceuticals, petroleum products, foods, and wastewater
The stainless steel probe can be used in liquids, semi-solids, and air/gas. Provides accurate readings when probe is immersed only 8mm (5/16"). Probe diameter: 3mm (1/8"). Probe stem length: 15.9 cm (61/4"). Overall probe length: 22.9 cm (9"). Thermometers feature one-button self-calibration. Display case is made of high-impact, chemical-resistant ABS plastic.
Display simultaneously shows maximum, minimum, and current probe temperatures, as well as room temperature. Alarm feature provides continuous visual and audio alerts when temperature rises above or falls below a programmable set point, even if temperature returns to non-alarm range.
Specifications
- Catalog No:
- 89094-758
- 89094-756
- 89094-754
- 89094-752
- Temperature resolution:
- 0.01°
- 0.01°
- 0.01°
- 0.01°
- W×D×H:
- 70×108×19 mm (23/4 × 41/4 × 3/4")
- 70×108×19 mm (23/4 × 41/4 × 3/4")
- 70×108×19 mm (23/4 × 41/4 × 3/4")
- 70×108×19 mm (23/4 × 41/4 × 3/4")
- Weight:
- 141.7 g (5 oz.)
- 141.7 g (5 oz.)
- 141.7 g (5 oz.)
- 141.7 g (5 oz.)