Spécifications
- Description:Waterproof Thermometer
- Temperature accuracy:±0.3% of reading plus 1°C (2°F) for –50 to 1000°C (–58 to 1832°F); ±0.5% of reading plus 1°C (2°F) for All Other Temperatures
- Temperature resolution:0.1° from –200 to 650°C, 0.1° from –328 to 999°F, 1° for all other temperatures
- W×D×H:7.6×3.2×18.4 cm (3×11/4 × 71/4")
- Weight:226.8 g (8 oz.)
- Range:–200...1300 °C (–328...2372 °F)
Spécifications
A propos de cet article
Wide-range thermometer has shock-resistant ABS plastic casing.
Wide-range thermometer has shock-resistant ABS plastic casing. Waterproof design stops dirt, dust, fumes, and water, making it ideal in harsh lab or plant environments. Features include °C or °F display, minimum and maximum temperature recall, differential temperature from a set point, hold button to capture current reading, and backlight button. The microprocessor updates the display 2.5 times per second. The 1.6cm (5/8") digits are readable from 3m (10').
- Traceable
- Temperature in °C and °F
- Waterproof design
- Highly visible display
- Minimum and maximum temperature recall
Waterproof design stops dirt, dust, fumes, and water, making it ideal in harsh lab or plant environments. Features include °C or °F display, minimum and maximum temperature recall, differential temperature from a set point, hold button to capture current reading, and backlight button. The microprocessor updates the display 2.5 times per second. The 1.6 cm (5/8") digits are readable from 3m (10').