Stands for Heidolph® Hei Torque Overhead Stirrers, Heidolph
Fournisseur: Heidolph NA, LLC
Certificats
A propos de cet article
Designed for use with the Heidolph Hei Torque series of overhead stirrers. These stands provide sturdy support for stirrers during mixing procedures.
The Universal stand S2 has a length of 700mm. The Universal stand S2 XXL has a length of 1000mm. The Telescope stand provides vertical displacement of 725–1025mm. All stands feature an H-shaped base measuring 425x420mm. Fastening clamps are not included.
Customers Who Bought This Also Bought