Stainless Steel Worktables, Straight Edge, Bandy
Supplier: Bandy
Certificates
About this item
These 16-gauge, type 304 stainless steel tables provide a sturdy work area in the cleanroom.
- Number 4 finish
- Working height: 76.2 cm (30")
- Straight front and rear edges
Table legs are round with a Ø of 4.1 cm (1⁵/₈"). Tables are available with a perforated or solid top.
Ordering information: Shipped assembled.
Customers Who Bought This Also Bought