Cupsink and Turret Kit for Basic™ Laboratory Hoods, Labconco®
# de catalogue 15202-652
Fournisseur: Labconco
Product available on GSA Advantage®
Spécifications
- Description:Cupsink and Turret Kit
- Cat. no.:15202-652
- Supplier No.:4852500
Spécifications
A propos de cet article
Kit includes a 7.6x15.2cm (3x6") polypropylene cupsink and chrome turret
The cupsink and turret mount on the right side wall. Plumbing pipe is not included.
Customers Who Bought This Also Bought