VWR® Magnetic Rare Earth Stir Bars
Fournisseur: VWR International
Certificats
A propos de cet article
Molded in pure, FDA-approved PTFE, these samarium-cobalt elliptical magnets feature increased magnetic strength and are almost totally resistant to demagnetization.
- Autoclavable
- PTFE construction
- Highly resistant to demagnetization
Rare Earth magnets are identified by a chemically inert carbon black spot.
Customers Who Bought This Also Bought