Barnstead B-pure™ Cartridge Water Purification System, Thermo Scientific
Fournisseur: Thermo Fisher Scientific
CE Compliant
Certificats
A propos de cet article
Thermo Scientific™ B-Pure Water Purification System is customizable for all pretreatment or deionization needs
Modular design provides the ability to add additional holders as needed. Quarter-turn, quick-release canisters make cartridge changes easy. All models accept Thermo Scientific Pura-Lite™ indicator to monitor water quality.
Pura-Lite™ indicators are easy-to-read lights. Green light indicates acceptable water purity, and red light indicates unacceptable water purity. Cell thread is 12.7 mm (1/2") NPTM.
B-Pure water purification system is an economical, laboratory-grade water system that can be customized to meet specific application needs. Recommended applications include pretreatment and deionization.
Ordering information: Canisters sold separately.
Spécifications
- Catalog No:
- 26302-250
- 82002-642
- 26302-448
- 26302-253
- Max. flow rate:
- 4 L per minute
- 4 L per minute
- 4 L per minute
- 4 L per minute
- Max. inlet pressure:
- 100 psig
- 100 psig
- 100 psig
- 100 psig
- Feed water:
- 4 to 49°C (40 to 120°F)
- 4 to 49°C (40 to 120°F)
- 4 to 49°C (40 to 120°F)
- 4 to 49°C (40 to 120°F)
- Inlet/outlet connection:
- 12.7 mm (1/2") NPTF
- 12.7 mm (1/2") NPTF
- 12.7 mm (1/2") NPTF
- 12.7 mm (1/2") NPTF