VWR® Spinbar® Magnetic Stir Bars, Saturn
Fournisseur: VWR International
Certificats
A propos de cet article
Many magnetic stirring bars can be impeded by powders in a liquid, but the Saturn Spinbar® Magnetic Stirring Bar rises to this stirring challenge.
- Large sphere at the bar’s center lifts the stirring bar ends while spinning
- Reduced surface contact area allows the bar to spin with decreased drag
- Alnico magnet encapsulated in FDA Grade, USP Class VI Teflon® PTFE low-friction coating; durable and inert
- Manufactured in a registered ISO 9001:2008 facility, as verified by SGS certification
- Autoclavable at 121°C (250°F)
Magnetic stir bars are available with a length of 40 or 50mm (1.5 or 2") to match with a wide variety of vessels with both round and flat bottoms.
Customers Who Bought This Also Bought