VWR® Magnetic Stir Bar Retrievers, PTFE
Fournisseur: VWR International
Certificats
A propos de cet article
Molded in pure, FDA-approved PTFE, these retrievers are inert and non-contaminating.
- Inert
- PTFE fabrication
- Autoclavable
- Magnetic cores available in different variants
Available with standard Alnico V or samarium cobalt rare earth magnetic core. Autoclavable.
Customers Who Bought This Also Bought