VWR® Spinbar® Disposable Stir Bars, Octagon
Fournisseur: VWR International
Certificats
A propos de cet article
The multifaceted stirring bar surface adds turbulence relative to similar size smooth cylindrical stirring bars.
- Ideal for single use applications such as mixing radioactive reagents
- Disposable; eliminates potential for cross-contamination
- Alnico magnet encapsulated in FDA grade Teflon® PTFE is durable and inert
- Autoclavable
For single-use application where efficiency and cross-contamination are a concern. Alnico V magnet.
Customers Who Bought This Also Bought