SP Bel-Art Spinbar® Combinatorial Chemistry Stir Bars, Bel-Art Products, a part of SP
Catalog # 80086-316
Supplier: Bel-Art Products, a Part of SP
Specifications
- Coating:PTFE
- Ø:5 mm (3/16")
- Description:Flea Micro Spinbar® Stir Bars
- Length:2 mm (3/32")
- Cat. no.:80086-316
- Shape:Egg-shaped
- Supplier No.:371199009
Specifications
About this item
Spinbar® magnetic stirring bars are suitable for combinatorial chemistry applicaitons.
- Well suited for use in round bottom flasks
- Bulk packaging for use with multiple reactions
- Teflon®-coated
The Flea Micro Spinbars® are Teflon® coated for use with organic solvents. Bulk packaging saves time, eliminating individual blister packs.
Customers Who Bought This Also Bought