VWR® Spinfin® Double Stir Bars
Fournisseur: VWR International
Certificats
A propos de cet article
Double Spinfin® Magnetic Stirring Bars agitate liquids in round bottom vessels such as test tubes, cylinders, and flasks, where other stirring bars cannot be used.
- Fins on top and bottom ensure complete agitation
- Maintains its position in a vessel and will not wobble
- Alnico magnet encapsulated in FDA Grade, USP Class VI Teflon® PTFE low-friction coating; durable and inert
- Autoclavable
Manufactured in a registered ISO 9001:2008 facility, as verified by SGS certification.
Customers Who Bought This Also Bought