VWR® Spinfin® Magnetic Stir Bars
Fournisseur: VWR International
Certificats
A propos de cet article
Spinfin® magnetic stirring bars agitate liquids in round bottom vessels such as test tubes, cylinders and flasks, where other stirring bars cannot be used
- Maintains its position in a vessel and will not wobble
- Manufactured in a registered ISO 9001:2008 facility, as verified by SGS certification
- Autoclave safe
Alnico magnet encapsulated in FDA grade Teflon® PTFE is durable and inert.
Customers Who Bought This Also Bought