Synthware Vacuum Manifold with PTFE Stopcocks, Kemtech America
Fournisseur: Kemtech America
CE Compliant
Certificats
A propos de cet article
General purpose manifolds are available with either 2 mm or 4 mm PTFE stopcocks
- Heavy wall borosilicate construction
- Teflon® stopcocks
- 4 or 6 port options available
The main tube has a 22 mm O.D., and features a serrated hose connection (10 mm O.D.) at the right, left, or both right and left sides. Delivery ports have 10 mm serrated hose connections. Distance between stopcocks is 100 mm.
Customers Who Bought This Also Bought