Synthware Storage Flask with Outer Joint, Kemtech America
Fournisseur: Kemtech America
CE Compliant
Certificats
A propos de cet article
Storage flasks are equipped with a high vacuum valve that features a PTFE O-ring at the tip.
- Hi vacuum valves
- Externally threaded
- Heavy wall borosilicate construction
- Ground glass joints
Our externally threaded storage tube is designed for storage of air sensitive materials.
Customers Who Bought This Also Bought