Undercounter Laboratory Flammables Safety Cabinets, SECURALL®
Fournisseur: Securall Cabinets
Certificats
A propos de cet article
Designed for undercounter placement, these laboratory cabinets feature a kick recess making it comfortable to stand at the work area.
- Self-close cabinets feature a fusible link that melts at 71 °C (165 °F) allowing the doors to automatically close and latch
- All cabinets have a large 'FLAMMABLE – KEEP FIRE AWAY' label
Cabinets are constructed of double-wall 18-gauge galvanized steel and have 5.1 cm (2") vents with flash arrestors for easy fume hood installation. Additional features include flush-mounted locking handle, three-point self-latching closure, one adjustable 18-gauge galvanized steel shelf, and full-length heavy-gauge piano hinges.
Certifications: Cabinets meet NFPA Code 30 specifications. Manual close cabinets are FM approved.
Spécifications
- Catalog No:
- 15001-702
- 15001-700
- 15001-704
- 15001-706
- 15001-698
- 15001-696
- Environmentally Preferable:
- Sustainable Materials
- Sustainable Materials
- Sustainable Materials
- Sustainable Materials
- Sustainable Materials
- Sustainable Materials
- Environmental certification:
- SMaRT Certified
- SMaRT Certified
- SMaRT Certified
- SMaRT Certified
- SMaRT Certified
- SMaRT Certified