Accessories for Heidolph® Hei Flow Peristaltic Pump Systems
Fournisseur: Heidolph
Certificats
A propos de cet article
Spécifications
- Catalog No:
- CA11009-640
- CA11009-636
- CA11010-276
- CA11009-638
- CA11009-632
- CA11009-634
- CA11009-630
- CA11009-624
- CA11009-626
- CA11009-642
- CA11009-644
- Flow rate:
- 0.005–40.8 mL/min.
- 0.6–336 mL/min.
- 0.5–40.8 mL/min.
- 1–3644 mL/min.
- 0.01–8.2 mL/min.
- 1–3644 mL/min.
- 0.7–2217 mL/min.
- 23.3–3300 mL/min.
- 0.29–25.2 mL/min.
- 1.0–336 mL/min.