Black Phenolic Screw Caps, PolyCone Liner, Qorpak®
Fournisseur: Qorpak
Certificats
A propos de cet article
PolyCone lined caps form an exceptionally tight seal and offer a good chemical barrier
These black phenolic screw caps, polyCone liner are made of phenolic resin.
This liner molds itself around the sealing areas of the finish as the cap is screwed on, virtually eliminating leakage, evaporation, contamination, binding and backing off. Great for everything from liquids, to soaps, to macro-invertebrates.
Customers Who Bought This Also Bought