Black Phenolic Screw Caps, Pulp/Tinfoil Liner, Qorpak®
Fournisseur: Avantor
Certificats
A propos de cet article
Pulp/Tin Foil lined caps offer good chemical resistance and are typically used with solids and powders
These black phenolic screw caps are made of phenolic resin.
Good resistance to hydro-carbons, alcohols, ketones and oils. Not
Customers Who Bought This Also Bought