Corrosive Storage Cabinets, SECURALL®
Fournisseur: Securall Cabinets
Certificats
A propos de cet article
Cabinet features adjustable shelving and leveling legs, three-point latching system, 5.1cm (2") vents with flash arrestors, flush-mounted locking handle, a sliding door, and a 5.1cm (2") leakproof sill
Caution: Not recommended for the storage of nitric or sulfuric acids.
Made with 18-gauge, galvanized steel with a double wall and a 3.8cm (11/2") air space. Shelves are equipped with removable corrosive-resistant polyethylene trays to contain spills and finished with two coats of corrosive-resistant blue epoxy enamel paint. Large warning label reads “CAUTION CORROSIVES.”
Certifications: Cabinets comply with OSHA and NFPA Code 30 requirements. All cabinets, with the exception of 14227-648, are FM approved.
Spécifications
- Catalog No:
- 14227-648
- 15001-690
- 15001-688
- 15001-686
- 14226-894
- 15001-694
- 14226-892
- 15001-692
- Environmentally Preferable:
- Sustainable Materials
- Sustainable Materials
- Sustainable Materials
- Sustainable Materials
- Sustainable Materials
- Sustainable Materials
- Sustainable Materials
- Sustainable Materials
- Environmental certification:
- SMaRT Certified
- SMaRT Certified
- SMaRT Certified
- SMaRT Certified
- SMaRT Certified
- SMaRT Certified
- SMaRT Certified
- SMaRT Certified